Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Tubing Clamp, Ace Glass Incorporated
Supplier: Ace Glass
11145-20
89075-638BX 128.17 USD InStock
89075-638
Tubing Clamp, Ace Glass Incorporated
Clamps Tubing and Sanitary Clamps
This tubing clamp eliminates the older method of wiring rubber tubing to glass
Simple in design and easy to use. Requires only one hand to apply; can be instantly applied or removed. Fits standard 10–12mm fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty