Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Fiberglass Support Rods, Ace Glass
Supplier: Ace Glass
11167-15 11167-11 11167-46 11167-40 11167-44 11167-05 11167-32 11167-09 11167-42 11167-36 11167-07 11167-38 11167-03 11167-13
89075-670EA 22.47 USD InStock
89075-670 89075-672 89075-660 89075-674 89075-652 89075-662 89075-676 89075-654 89075-664 89075-678 89075-656 89075-666 89075-658 89075-668
Fiberglass Support Rods, Ace Glass
Support Rods and Frames
These Fiberglass Support Rods are used in place of aluminum rods that pit or become unsightly when used in corrosive atmosphere
Off-white or black color of these rods lends a more attractive appearance to lab setups. For use with all standard laboratory connectors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty