Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Stainless Steel Support Rods, Ace Glass
Supplier: Ace Glass
11178-10 11178-04 11178-14 11178-12 11178-06 11178-16 11178-08
89075-694EA 130.43 USD InStock
89075-694 89075-696 89075-698 89075-686 89075-688 89075-690 89075-692
Stainless Steel Support Rods, Ace Glass
Support Rods and Frames
303 stainless steel support rods, for use in place of aluminum or fiberglass
For use with all standard laboratory connectors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty