Would you like to log in again?
High Temperature Graphite Ferrules, Ace Glass
Supplier: Ace Glass
11720-20 11720-12 11720-18 11720-16
89075-760PK 131.47 USD InStock
89075-760 89075-762 89075-764 89075-758
High Temperature Graphite Ferrules, Ace Glass
Fittings and Connectors
These single piece, high purity graphite ferrules require no back ferrule, and no organic binders are used
They are ideal for glass-to-glass or glass-to-metal connections. All sizes are used with a #7 Ace-Thred. Due to their high purity and unique properties, these ferrules make a mono-molecular seal — one of the strongest seals known.
Ordering information: Note that ferrules are not always suitable for vacuum applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty