Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mantles, Fabric/Cloth, Ace Glass Incorporated

Supplier: Ace Glass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

12036-17 12036-24 12036-21 12036-23 12036-19
89075-880EA 849.97 USD InStock
89075-880 89075-882 89075-884 89075-886 89075-888
Mantles, Fabric/Cloth, Ace Glass Incorporated
Heaters Heating Mantles
Heating mantles are designed for use with reaction flasks

Upper temperature limit is 450°C (842°F). A 122cm (4') cord and locking connector are included. Units must be operated through a temperature controller.

Certifications: CSA approved.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty