Would you like to log in again?
Mantle for Squibb-Type Funnels, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
12038-04 12038-08 12038-06 12038-10
89075-918EA 1435.9 USD InStock
89075-918 89075-912 89075-914 89075-916
Mantle for Squibb-Type Funnels, Ace Glass Incorporated
Heaters Heating Mantles
Designed for use with Squibb type separatory funnels
Mantles are supplied with separable 122 cm (4'), 2-wire cord, and locking connector. Large sizes of one liter and up have viewing slots, as shown.
Certifications: CSA approved.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty