Would you like to log in again?
Mantle, Aluminum Housing, Round Bottom, Ace Glass Incorporated
Supplier: Ace Glass
12043-19 12043-25 12043-21 12043-31 12043-32 12043-17 12043-27 12043-05 12043-13 12043-23 12043-33 12043-29 12043-07
89076-020EA 630.52 USD InStock
89076-020 89075-962 89075-964 89075-952 89076-018 89075-966 89075-954 89075-968 89075-956 89076-024 89076-034 89076-022 89076-032
Mantle, Aluminum Housing, Round Bottom, Ace Glass Incorporated
Heaters Heating Mantles
This mantle offers the benefits of grounding via its aluminum housing
The heating element is embedded in layers of glass fabric to protect the flask wall from thermal strain, and the mantle is thoroughly covered with glass insulation to prevent heat from being radiated outward. The glass fabric affords operating temperatures to 450°C (842°F). Mantle is CSA certified.
Ordering information: Mantle is supplied complete with detachable 122cm (4'), three-wire cord and locking connector. An appropriate size support (see 89076-334) is required, but not included. Mantle must be operated through a temperature controller. A 230V version is also available for 250mL and larger mantles. Contact your Avantor representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty