Would you like to log in again?
Glas-Col® Low Profile Aluminum Housed Heating Mantle, Ace Glass
Supplier: Ace Glass
Glas-Col®
12050-38 12050-41 12050-43 12050-45 12050-34 12050-36
89076-008EA 4119.77 USD InStock
89076-008 89076-006 89076-016 89076-014 89076-012 89076-010
Glas-Col® Low Profile Aluminum Housed Heating Mantle, Ace Glass
Heaters Heating Mantles
This mantle is used with reaction assemblies and is CSA certified
Easily fits into ACE 4-post reactor stands. Temperature range is Ambient 10°C to 450°C. Must be operated through a temperature controller. With detachable 4' cord and locking connector. Available with or without bottom opening.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty