Would you like to log in again?
Resin and Reaction Flask Mantle, Aluminum Housing, Cylindrical Flasks, Ace Glass Incorporated
Supplier: Ace Glass
12058-33 12058-30 12058-03 12058-07
89076-092EA 2235.77 USD InStock
89076-092 89076-090 89076-078 89076-076
Resin and Reaction Flask Mantle, Aluminum Housing, Cylindrical Flasks, Ace Glass Incorporated
Heaters Heating Mantles
This resin and reaction flask mantle is designed mainly for use with pressure reactors, although it will fit many other cylindrical reaction flasks as well
This mantle is designed to work with flasks such as 89054-946, 89055-050, and 89055-132 (among others) with appropriate O.D. The 89076-076 mantle is designed for flasks with a flat bottom and rounded corners, such as 89055-058.
The mantle is aluminum housed with a solid bottom. It offers a temperature range of ambient plus 10 to 450°C (842°F).
Certifications: CSA approved.
Ordering information: Mantle includes 122cm (4') detachable cord with locking connector. Mantle must be operated through a temperature controller. For more information on matching this mantle with appropriate flask(s), contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty