Would you like to log in again?
Resin and Reaction Flask Mantle, Aluminum Housing, Bottom Opening, Ace Glass Incorporated
Supplier: Ace Glass
12058-47 12058-53 12058-49 12058-44 12058-51 12058-55
89076-096EA 1567.79 USD InStock
89076-096 89076-094 89076-104 89076-102 89076-100 89076-098
Resin and Reaction Flask Mantle, Aluminum Housing, Bottom Opening, Ace Glass Incorporated
Heaters Heating Mantles
Resin and reaction flask mantle is designed mainly for use with pressure reactors with a bottom valve or outlet
Mantle 89076-094 is designed for flasks having a flat bottom with rounded corners, such as 89054-406.
These mantles also fit many other cylindrical reaction flasks, such as 89083-812, 89055-162, 89055-122 and 89083-998 (among others) with appropriate O.D. Mantles have an aluminum housing and a bottom opening. Temperature range is ambient plus 10 to 450°C (842°F).
Certifications: CSA approved.
Ordering information: Mantle includes 122cm (4') detachable cord with locking connector. Mantle must be operated through a temperature controller. For more information on matching this mantle with appropriate flask(s), contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty