Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Type J Thermocouple Probe, Ace Glass Incorporated

Supplier: Ace Glass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

12110-09 12110-24 12110-17 12110-26 12110-15 12110-25 12110-27
89076-448EA 227.7 USD InStock
89076-448 89076-458 89076-446 89076-456 89076-454 89076-452 89076-450
Type J Thermocouple Probe, Ace Glass Incorporated
Electrodes pH Electrodes
Type J temperature probes provide economical measurement from -20° C to 550° C

Probes available sheathed with stainless steel, inconel or simply insulated wire. With male mini plug lead.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty