Would you like to log in again?
Thermocouple Probe with Permanently Attached Leads, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
12140-06 12140-57 12140-37 12140-28 12140-16 12140-61 12140-25 12140-39 12140-05 12140-21 12140-53 12140-10 12140-59 12140-46 12140-20 12140-19 12140-04 12140-17 12140-14 12140-41 12140-18 12140-36 12140-62 12140-26 12140-48 12140-58 12140-45 12140-51
89076-570EA 343.87 USD InStock
89076-570 89076-592 89076-590 89076-618 89076-616 89076-614 89076-612 89076-578 89076-610 89076-598 89076-576 89076-574 89076-596 89076-572 89076-594 89076-580 89076-608 89076-606 89076-604 89076-602 89076-568 89076-600 89076-622 89076-588 89076-620 89076-586 89076-584 89076-582
Thermocouple Probe with Permanently Attached Leads, Ace Glass Incorporated
Electrodes pH Electrodes
Type J or Type K thermocouple sensors with plain Inconel or PTFE-coated sheath, 12' PTFE-covered permanently attached leads with stainless steel overbraid that prevents kinking, and subminiature male connector ends
  • Lead Attachment Permanent
Interface between sensor and lead has a spring transition for strain release. Plain Inconel sheath has a temperature range of -60 to 550°C. PTFE-coated sheath is limited to 200°C over long periods, 230°C maximum for short periods. Available in a variety of sheath lengths and O.D.'s
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty