Would you like to log in again?
Stoppers, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
12634-24 12634-20
89076-996PK 289.49 USD InStock
89076-996 89076-994
Stoppers, PTFE, Ace Glass Incorporated
Stoppers PE/LDPE/HDPE Stoppers
Virgin TFE/PTFE stoppers with built-in black Delrin stopper extractor ring
Eliminates the hazard of removing a frozen stopper from glass vessel. You need only turn red nut clockwise to exert a gentle force against the vessel’s lip and the non-stick stopper is extracted.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty