Would you like to log in again?
Universal Tubing Connector, Ace Glass Incorporated
Supplier: Ace Glass
12723-12
89077-442PK 287.19 USD InStock
89077-442
Universal Tubing Connector, Ace Glass Incorporated
Fittings and Connectors
This connector permits flangeless instant connection
The larger end accepts tubing over O.D. range 4 to 11 mm. The smaller end accepts tubing over O.D. range 0.5 to 4 mm. Large cap is Delrin and the small cap is 1/4" threaded polypropylene PTFE cones for all-PTFE connections are included. PTFE/Tefzel Caps are available for use at temperatures to 220°C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty