Would you like to log in again?
Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
12770-59 12770-12 12770-14 12770-28 12770-52 12770-31 12770-54 12770-26 12770-32 12770-18 12770-38 12770-24 12770-16 12770-30 12770-27 12770-36
89077-528EA 66.17 USD InStock
89077-528 89077-506 89077-516 89077-508 89077-518 89077-502 89077-524 89077-512 89077-504 89077-526 89077-514 89077-520 89077-530 89077-522 89077-532 89077-510
Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
Fittings and Connectors
These all-PTFE fittings have a male NPT (MNPT) thread at one end and either tube compression, a hose barb, or MNPT thread at the other end.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty