Would you like to log in again?
Septa for Serum Bottles, Ace Glass Incorporated
Supplier: Ace Glass
12908-60
89077-548PK 147.58 USD InStock
89077-548
Septa for Serum Bottles, Ace Glass Incorporated
Stoppers Rubber Stoppers
Septa, white silicone with structurally bonded PTFE facing
Resists syringe coring. For 20mm mouth O.D., these septa have an O.D. of 19mm, and they are 3mm thick. Use with aluminum crimp seals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty