Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Arrow Laboratory Stirrers, Ace Glass Incorporated
Supplier: Ace Glass
13544-20 13542-04 13542-02 13542-25 13543-12
89077-726EA 1396.97 USD InStock
89077-726 10767-212 89077-724 10767-210 89077-720
Arrow Laboratory Stirrers, Ace Glass Incorporated
Stirrers Overhead Stirrers
Arrow high torque, variable speed laboratory stirrers are designed to handle a wide variety of laboratory applications
Supplied with a 10" long, 9/16" diameter support rod. Motor shaft is 3/8" (9.5 mm) in diameter. Stand base fabricated of steel with 5/8" O.D. x 29" long stainless steel support rod.
Ordering information: Stirrer shaft, propeller, and coupling are for use with Models 850, 6000 and 1750. For clamp, see Ace Glass' power hold clamp to use with the support stand.
Mounting rod for ease in attaching to a laboratory lattice or support stand
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty