Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Laboratory Stirrers, Ace Glass Incorporated
Supplier: Ace Glass
13649-07 13649-09 13649-19
89077-804EA 1174 USD InStock
89077-804 89077-806 89077-808
Laboratory Stirrers, Ace Glass Incorporated
Stirrers Overhead Stirrers
Totally enclosed, 1/40hp DC permanent magnet motor with dual 8 mm (5/16") armature and gear shafts with milled flats
Motor is supplied with 1.5 meter three-wire cable with plug and ground lead for connection to controller. Measures: 5-1/2" x 4" x 4". Weight: 4 lbs. 7 oz. Controller is solid state 120 V, 10 amp AC maximum or 0–120 V, 6 amp DC maximum. Features rear ring stand clamp, Forward-Off-Reverse switch. Supplied with heavy duty 1.8 meter three-wire power cord with NEMA plug. Supplied with one-amp fuse to protect motor. Controller measures 102 (4") x 54 mm (2-1/8") x 41 mm (1-5/8"). It weighs 4.1 lbs. Complete consists of motor, controller and mounting rod.
Ace Glass Laboratory Stirrers Accessory Kit Includes (1) Each of the following:
89077-810
89077-812
89077-816
89077-818
89077-820
Armature shaft with maximum speed of 4000rpm.; 18:1 ratio gear shaft rated 4.2Kg.cm torque, up to 333rpm. Motor supplied with 1.5 meter 3-wire cable with plug and ground lead for connection to controller. Controller 89077-716: solid state 120v, 10 amp AC max. or 0-120v, 6 amp DC max. Features rear ring stand clamp, Fwd-Off-Rev. switch. Supplied with 1amp fuse and heavy duty 1.8 meter 3-wire power cord with NEMA plug
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty