Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
High Torque Economy Laboratory Stirrers, Ace Glass
Supplier: Ace Glass
13650-40 13650-12 13650-23
89077-824EA 744.48 USD InStock
89077-824 89077-826 89077-828
High Torque Economy Laboratory Stirrers, Ace Glass
Stirrers Overhead Stirrers
Motor and controller, motor is non-reversible, 1/12 hp DC, 5/16" dia shaft 1" long
stand shown in image is sold separately.
Recommended for use with 8081 Flexible Shaft or general stirring. Motor is adjustable from 280 - 2800 rpm and generates 60 oz/inch of nominal torque. Motor can only be used with the 120 V, 50/60Hz controller and is supplied with a 5/8" diameter mounting rod
Ordering information: Ace Glass Laboratory Stirrers, High Torque, Economy Complete Package Includes (1) Each:
89077-824
89077-826
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty