Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Stirrer Magnets with Raised Ring, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13655-29 13655-31 13655-33 13655-37 13655-25 13655-41 13655-44 13655-39
89077-876EA 66.85 USD InStock
89077-876 89077-886 89077-878 89077-872 89077-882 89077-874 89077-884 89077-880
Stirrer Magnets with Raised Ring, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
Octagonal-shaped, PTFE stirrer magnets feature a large 25mm (1") pivot ring (PTFE collar).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty