Would you like to log in again?
Stirrer Magnets, Octogonal, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13657-59 13657-51 13657-69 13657-09 13657-49 13657-17 13657-05 13657-75 13657-61 13657-31 13657-25 13657-63 13657-23 13657-33 13657-57 13657-35 13657-37 13657-11 13657-07 13657-43 13657-76
89077-924EA 20.38 USD InStock
89077-924 89077-946 89077-956 89077-948 89077-926 89077-936 89077-958 89077-920 89077-942 89077-964 89077-930 89077-952 89077-922 89077-944 89077-954 89077-932 89077-960 89077-962 89077-940 89077-928 89077-938
Stirrer Magnets, Octogonal, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
This octagonal PTFE stirrer magnet, available in a wide variety of lengths and diameters, features a center pivot ring
Best for use in containers with bases that are slightly curved or uneven. Stirrer magnets are available in red, yellow, or blue for easy and instant identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty