Would you like to log in again?
Stirrer Magnets, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13666-06 13666-08 13666-05 13666-12 13666-04 13666-10
89077-996EA 33.45 USD InStock
89077-996 89077-998 89078-000 89077-992 89077-994 89078-002
Stirrer Magnets, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
These PTFE stirrer magnets are designed specifically for use in round-bottom test tubes and flasks
The 9x6mm size (only) is designed especially for use with 10mm standard cuvettes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty