Would you like to log in again?
Needles, Standard Hypodermic, Ace Glass Incorporated
Supplier: Ace Glass
13683-32 13683-29 13683-23
89078-054PK 676.84 USD InStock
89078-054 89078-058 89078-056
Needles, Standard Hypodermic, Ace Glass Incorporated
Syringe Needles
Standard hypodermic stainless steel needles with 12° regular medical point tip and female Luer-Lok hub
Packed 12 needles on card-cellophane wrapped. Each card is individually boxed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty