Would you like to log in again?
Needles, Special Length, Ace Glass Incorporated
Supplier: Ace Glass
13684-19 13684-15 13684-07 13684-11
89078-061CS 1280.15 USD InStock
89078-061 89078-065 89078-063 89078-067
Needles, Special Length, Ace Glass Incorporated
Syringe Needles
Special length 304 stainless steel needles with a deflected septum point and standard female hub
The deflected septum point is equivalent to B-D Huber or Hamilton Style 1 and 2.
Packaging: Packed 6 per package or a case of 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty