Would you like to log in again?
Hyvac Light Pump Oil, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
14035-09 14035-06 14035-21
89078-204EA 482.61 USD InStock
89078-204 89078-202 89078-200
Hyvac Light Pump Oil, Ace Glass
Lubricants
Hyvac Light Pump Oil incorporates two essential high vacuum characteristics: low vapor pressure which does not materially increase at temperatures up to 50 °C, and viscosity sufficiently low for use at 15 °C
It remains fairly constant up to 50 °C. For use with belt-driven or direct-drive pumps.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty