Would you like to log in again?
Flange, Duran®-Style, Ace Glass
Supplier: Ace Glass
15310-20 15311-23 15311-31 15310-25 15310-22 15310-15 15311-26 15311-21 15310-30 15311-16
89078-302EA 530.21 USD InStock
89078-302 89078-292 89078-290 89078-286 89078-296 89078-284 89078-294 89078-300 89078-288 89078-298
Flange, Duran®-Style, Ace Glass
Fittings and Connectors
This Duran®-style flange is used by glassblowers to fabricate reaction flasks.
It is available with or without an o-ring groove. For clamps, see 89055-114 (choose appropriate size). Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty