Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Model 8100 Variable Repetitive Syringe Dispenser, Nichiryo America
Supplier: Nichiryo
8100
89078-434EA 695.37 USD InStock
89078-434
Model 8100 Variable Repetitive Syringe Dispenser, Nichiryo America
Pipettes Repetitive Pipettors
The lightweight and slender Nichiryo 8100 syringe dispenser provides 35 methods of liquid dispensing throughout the 1 to 5000 µl volume range.
The ergonomic design permits easy handling without fatigue. Syringes are quickly and easily interchangeable, and volume setting is conveniently adjustable by dial. Dispensers offer excellent accuracy and reproducibility. With the exception of the 3 and 60 ml units, all syringes are autoclavable. Syringe 89078-442 includes a spacer in the nozzle specially designed for use with low liquid pressure microtiter plates. Syringe 89078-450 includes one adaptor to mount syringe to dispenser handle.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty