Would you like to log in again?
Model 8800 8-Channel Variable Repetitive Syringe Dispenser, Nichiryo America
Supplier: Nichiryo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
88-CB 88-SBR 88-TY 8800 88-CA 88-SAR
89078-490PK 323.93 USD InStock
89078-490 89078-494 89078-492 89078-488 89078-498 89078-496
Model 8800 8-Channel Variable Repetitive Syringe Dispenser, Nichiryo America
Pipettes Repetitive Pipettors
The 8800 Syringe Dispenser offers a unique approach to multichannel dispensing.
A choice of replaceable syringes mounted on interchangeable cartridges allow the user to dispense six different volumes of liquid (10, 20, 25, 40, 50, and 100µl) into a microtiter plate. Multiple dispensing can be achieved by repeatedly pressing the end button. The lightweight and slender design allows for easy handling without fatigue, even over extended periods of use. An intuitive adjustment dial provides quick volume identification. Syringes may be quickly and easily interchanged and replaced. Both syringes and cartridges are autoclavable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty