Would you like to log in again?
VWR® Inkjet / VWR® Inkjet Plus Microscope Slides, Premium
Supplier: VWR
VWR®
IJL-6100-4-601640 IJL-6-PS-4-601640 IJL6113P4601 IJL-2-PS-4-601640 IJL-1-PS-4-601640 IJL-7-PS-4-601640 IJL-6104-4-601640 IJL-9-PS-4-601640 IJL6115P4601
89078-525CS 937.81 USD InStock
89078-525 89078-535 89078-501 89078-523 89078-533 89078-527 89078-508 89078-519 89078-531
VWR® Inkjet / VWR® Inkjet Plus Microscope Slides, Premium
Slides Microscope Slides
Slides have a frosted tab which is designed to be directly printed upon by Leica® Microsystems IP S ink jet printer.
  • Available in various colors
  • Frosted edges
Ideal for histology, cytology, hematology, and microbiology applications. Frosted tabs are available in various colors. Inkjet Plus slides include a plus charge symbol.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty