Would you like to log in again?
KIMAX® Volumetric Flasks with Color-Coded [ST] PTFE Stopper, Wide Mouth, Class A, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
92812F-5 92812F-25 92812F-50 92812F-20 92812F-200 92812F-250 92812F-1000 92812F-100 92812F-10 92812F-2000 92812F-500
89079-180CS 371.58 USD InStock
89079-180 89079-190 89079-182 89079-192 89079-184 89079-194 89079-186 89079-174 89079-188 89079-176 89079-178
KIMAX® Volumetric Flasks with Color-Coded [ST] PTFE Stopper, Wide Mouth, Class A, Kimble Chase
Flasks Volumetric Flasks
Flasks feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty