Would you like to log in again?
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
92812P-50 92812P-20 92812P-200 92812P-5 92812P-250 92812P-500 92812P-2000 92812P-25 92812P-1000 92812P-100 92812P-10
89079-214CS 208.86 USD InStock
89079-214 89079-202 89079-216 89079-204 89079-206 89079-208 89079-196 89079-210 89079-198 89079-212 89079-200
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, Kimble Chase
Flasks Volumetric Flasks
Flasks feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty