Would you like to log in again?
KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, PTFE Stopper, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE® RAY-SORB®
92822F-500 92822F-2000 92822F-250 92822F-5 92822F-50 92822F-10 92822F-100 92822F-25 92822F-200 92822F-1000 92822F-20
89079-268CS 458.74 USD InStock
89079-268 89079-270 89079-280 89079-272 89079-282 89079-274 89079-262 89079-276 89079-264 89079-278 89079-266
KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, PTFE Stopper, DWK Life Sciences
Flasks Volumetric Flasks
Flasks are made of red-tinted glass that provides protection from UV light
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety. Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty