Would you like to log in again?
VIAL SS SLIP FIT 56X92MM
Supplier: CHEMPLEX INDUSTRIES
Stainless Steel SlipFit* SpectroVials*^ General purpose comminuting and blending vials. Use for grinding moderately hard sample materials. Interior ends are concaved and both halves are designed to easily slip on and off. Highly rust resistant grade of stainless steel. Length: 92mm. Dia: 56mm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL SS SLIP FIT 56X92MM 89079-782 1149 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty