Would you like to log in again?
SAMPLE CUP RESEAL 30.7MM PK100
Supplier: CHEMPLEX INDUSTRIES
Sample Cups^ Re-sealable XRF Sample Cups include ventable caps with overflow reservoir and integrated thin-film trimmer. Integrated serrated edged snap-on rings allow extraneous thin-film trimming on the assemble sample cup. Diameter: 30.7mm. Height: 23.5mm. Capacity: 7cc.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAMPLE CUP RESEAL 30.7MM PK100 89079-804 1330-SE Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty