Would you like to log in again?
SAMPLE CUP OVRFLW 31.2MM PK100
Supplier: CHEMPLEX INDUSTRIES
Sample Cups^ Double open-ended XRF Sample Cups feature new internal overflow reservoirs, snap-on covers and easy-to-use sleeves for thin-film sample support attachment. Outside diameter: 31.2mm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAMPLE CUP OVRFLW 31.2MM PK100 89079-818 2131 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty