Would you like to log in again?
PRESS 12 TON FORCE MANUAL
Supplier: CHEMPLEX INDUSTRIES
Manual 12 Ton Force Spectropress*^ Contemporary designed manual press delivers 12 tons of force. Compact and light weight for counter top and field operations. Accepts 13, 32, 35 and 40 mm stainless steel briquetting dies for XRF powdered sample briquetting and 13 mm dies for IR applications.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PRESS 12 TON FORCE MANUAL 89079-872 4312 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty