Would you like to log in again?
SpectroMicro® Sample Cups, Chemplex
Supplier: Chemplex
3110 3120
89079-910PK 132.69 USD InStock
89079-910 89079-908
SpectroMicro® Sample Cups, Chemplex
Sample Containers XRF Sample Cups
These sample cups Utilize FunnelShape™ technology that has a vented snap-On cap equalizes pressure differentials within the Cell and sample chamber to maintain a taut thin-film sample support plane.
  • Larger conically shaped upper portion of the funnel allows and facilitates sample introduction
  • Lower cylindrically shaped spout of the funnel represents the actual sample chamber
The collar is used to secure a thin-film sample support window substance to the Cell and designed for acceptance by 32 mm sample cup holders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty