Would you like to log in again?
Finnpipette™ F1 Variable Volume Pipettes, Thermo Scientific
Supplier: Thermo Scientific
Finnpipette™
4641010N 4641050N 4641130N 4641090N 4641040N 4641060N 4641020N 4641030N 4641080N 4641140N 4641140N 4641070N 4641040N 4641010N 4641120N 4641030N 4641090N 4641100N 4641110N 4641120N 4641070N 4641020N 4641110N 4641130N 4641050N
89080-433EA 565.18 USD InStock
CA89080-421 89080-433 89080-431 CA89126-245 CA89126-243 89080-419 89080-417 CA89080-569 CA89080-425 89080-415 89080-413 89080-423 CA89080-431 89080-421 89126-243 CA89080-419 89126-245 CA89080-417 89080-429 CA89080-415 89080-427 CA89080-413 89080-569 89080-425 CA89080-433
Finnpipette™ F1 Variable Volume Pipettes, Thermo Scientific
Pipettes Micropipettes
The enhanced volume adjustment on Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes is now silent and effortless with textured adjustment knob for a firmer feel when setting volumes.
  • Silent volume adjustment
  • Adjustable finger rest
  • Lightweight design
  • Secure volume lock
  • Increased air boost
  • Five-year warranty
The adjustable finger rest, and lightweight design maximizes comfort without sacrificing performance. Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase in air boost ensures efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.
Certifications: The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
Accessories information: Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
Ordering information: Five-year warranty with convenient online registration; may vary by country.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty