Would you like to log in again?
Axygen® Pipette Tip Reload System, Corning
Supplier: Corning
Axygen®
RFL-300-C RFL-222-C RFL-1200-C
89029-921CS 357.08 USD InStock
89029-921 89080-484 89029-919
Axygen® Pipette Tip Reload System, Corning
Pipette Tips
Gilson®-style and universal fit pipette tips in easy-to-load refill racks.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
Universal fit tips have a beveled reference and graduated marks.
Axygen® tip reload system contains durable refill rack that can be easily refilled by hand and is autoclavable. They are available in different colors and are packaged in bulk or racks.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty