Would you like to log in again?
Axygen® Automation Tips for Tecan®Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
TT-1000-CBK-HTR-S TT-200-CBK-HTR EVF-180-R-S TT-1000-CBK-HTR-24 TTF-20-C-HTR-S TTF-50-C-HTR-S EV-50-NTR TTF-1000-C-HTR-S EV-200-NTR-S EVF-205-WB-R-PB EV-50-R-S EV-50-R TT-1000-CBK-HTR TT-50-CBK-HTR EV-50-NTR-S TT-1000-C-HTR-S EVF-205-WB-R-S TT-50-CBK-HTR-S TTF-200-CBK-HTR-S EV-100-R TT-50-CBK-HTR-36 TT-20-CBK-HTR TT-200-CBK-HTR-S EV-200-NTR EV-200-NTR-S TT-200-C-HTR-S EV-50-NTR TTF-1000-CBK-HTR-S TT-200-C-HTR EVF-100-R-S EVF-50-R-S EV-200-R-S EV-200-NTR EV-200-R TTF-50-CBK-HTR-S EV-50-NTR-S TTF-20-CBK-HTR-S TT-1000-C-HTR TT-50-C-HTR TTF-200-C-HTR-S
10030-826CS 434.35 USD InStock
10030-826 89087-340 10030-828 89108-024 89080-498 89125-544 10030-822 89080-496 89080-494 10030-824 89040-104 89080-490 89040-102 89040-068 89040-084 89040-062 89040-060 89040-082 89040-100 89040-066 89040-064 89040-086 CA10030-828 89080-500 CA10030-826 CA10030-824 CA10030-822 89087-538 89040-058 89040-074 89040-096 89040-094 89040-072 89040-098 89080-506 89080-504 89040-092 89040-070 89040-090 89080-502
Axygen® Automation Tips for Tecan®Corning
Pipette Tips
PP, with or without filter, sterile or non sterile.
  • Black tips are conductive for liquid level-sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Packaging: Packaged in plastic blister packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty