Would you like to log in again?
Axygen® Automation Tips for PerkinElmer® Janus®/MultiProbe, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
RRP-50-C-R RRF-25-C-R-S RRF-175-CBK-R-S RRF-25-CBK-R-S RRP-200-CBK-R RRP-200-C-R-S RRF-175-C-R-S RRP-200-C-R RRP-50-CBK-R RRP-20-C-R
89080-544CS 902.25 USD InStock
89080-544 89080-554 89080-542 89080-552 89080-540 89080-550 89080-538 89124-954 89080-536 89080-546
Axygen® Automation Tips for PerkinElmer® Janus®/MultiProbe, Corning
Pipette Tips
Axygen® automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Engineered to the exact specifications
  • Complete protection against contamination
Axygen® offers a comprehensive line of tips for PerkinElmer® automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are certified RNase-/DNase-free and nonpyrogenic. Available in nonsterile and sterile formats. Clear and Conductive Black polypropylene tips are available in 96-well format designed for PerkinElmer® Janus®/MultiProbe Variospan-1, 4, 8 heads.
Certifications: Tips are certified RNase-/DNase-free, and nonpyrogenic.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty