Would you like to log in again?
ART™ Barrier Reload Insert Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific ART
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2789-RI 2739-05-RI 2769-RI 2739-RI 2749-RI 2769-05-RI 2779-RI 2749-05-RI 2779-05-RI
89082-142CS 1257.23 USD InStock
89082-142 89082-130 89082-140 89082-146 89082-134 89082-144 89082-132 89082-138 89082-136
ART™ Barrier Reload Insert Pipette Tips, Thermo Scientific
Pipette Tips
The ART™ (Aerosol Resistant Tip) Barrier Reload Insert pipette tips seal when exposed to potential contaminants, providing accurate and consistent performance.
The ART inserts are environmentally friendly. Individually packed blister tubs are made out of recycled plastic and are designed to be used with the reusable ART hinged racks - creating the ART Reload System. The pipette tips from the reload inserts clip securely into the hinged racks for efficient reloading.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty