Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Proline® Plus Single Channel Mechanical Pipettors, Sartorius
Supplier: Sartorius
Proline® Plus
728570 728565 728090 LH-728673 728060 728590 728040 728010 LH-728670 LH-728677 728545 728590 728020 728520 728580 728570 728575 LH-728674 728545 728515 728515 LH-728672 728050 728535 728030 728530 728090 728520 728030 LH-728671 728080 LH-728676 728580 728080 728567 728010 728550 728040 728060 728070 728535 728575 LH-728675 728550 728567 728020 728560 728560 728050 728530 728070 728565
75845-038EA 773.46 USD InStock
CA89082-274 CA89082-276 75845-038 89177-094 75845-040 CA89082-318 75845-044 75845-042 89082-304 CA89082-314 89082-302 CA89082-316 CA89082-310 89082-308 CA89082-278 89082-306 CA89082-312 CA89177-094 CA89082-308 CA89082-304 CA89082-306 CA89082-302 89082-284 89082-282 89082-322 89082-288 89082-320 89082-286 89082-280 89082-326 89082-324 89082-274 CA89082-284 CA89082-286 CA89082-320 CA89082-280 89082-278 89082-310 89082-276 CA89082-282 75845-048 75845-046 75845-052 75845-050 CA89082-326 89082-314 89082-312 CA89082-288 CA89082-322 89082-318 89082-316 CA89082-324
Proline® Plus Single Channel Mechanical Pipettors, Sartorius
Pipettes Micropipettes
Proline® Plus mechanical pipettors are designed to offer comfort and quality for everyday manual pipetting.
  • Ergonomic design with light pipetting forces, comfortable handle, and finger support for reduced risk of strain injuries
  • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure for heavier use
Pipettors offer good UV resistance and excellent chemical resistance. The devices are suitable for both left- and right-handed users. Thermal insulation of internal components ensures that the warmth of the hand does not affect pipetting results during extended use. Pipettors are fully autoclavable. To facilitate tip selection, color coding on the pipettor's display corresponds to color coded tip trays.
They combine durable construction with ease and lightness of use, and are therefore appropriate for liquid-handling professionals and students alike.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty