Would you like to log in again?
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
Supplier: Sorenson BioScience
OneTouch™
10350 10460 10320 10380 10400 10420
89082-350CS 827.87 USD InStock
89082-350 89082-344 89082-354 89082-363 89082-358 89082-338
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
Pipette Tips
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
  • Dual-shot tip design
  • Easy-to-seal
  • Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty