Would you like to log in again?
Pipette Tips, Sorenson BioScience
Supplier: Sorenson BioScience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
30140 30800T 30920T 30430 27120 30800 30170 23570 30460 34750 23580 30160 30920 30930 34770
15001-099CS 313.47 USD InStock
15001-099 89082-382 89082-376 89082-386 89082-374 14214-395 53550-073 89082-380 53550-067 89082-390 13503-464 53550-071 14006-902 14006-914 89082-378
Pipette Tips, Sorenson BioScience
Pipette Tips
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty