Would you like to log in again?
Adapter, Connecting, Ace Glass Incorporated
Supplier: Ace Glass
5129-25 5129-15 5129-11 5129-07
89082-508EA 133.48 USD InStock
89082-508 89082-510 89082-506 89082-504
Adapter, Connecting, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is used with vacuum manifolds as port connections.
  • O-ring joint on one end
  • Ace-Thred on opposite end
  • One o-ring included
  • Fabricated from borosilicate glass
Featuring a #15 o-ring joint on one end with an Ace-Thred on the opposite end. Supplied with one o-ring.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty