Would you like to log in again?
Adapter, Outer Socket Joint, Ace Glass Incorporated
Supplier: Ace Glass
5221-24 5221-28 5221-09 5221-20 5221-11 5221-05
89082-574EA 81.13 USD InStock
89082-574 89082-552 89082-550 89082-578 89082-576 89082-554
Adapter, Outer Socket Joint, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This series of adapters is designed for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
  • Outer socket joints
  • Various Ace-Thred and tube ends
  • Fabricated from borosilicate glass
Adapters feature 28/15 and 35/25 sockets with various Ace-Thred and tube ends.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty