Would you like to log in again?
O-Ring Joint to Standard Taper Inner Joint with Frit Connecting Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5117-18 5117-21
89082-622EA 119.79 USD InStock
89082-622 89082-620
O-Ring Joint to Standard Taper Inner Joint with Frit Connecting Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is used with vacuum manifolds as port connections.
  • O-ring joint on one end
  • Glass frit below o-ring joint
  • Inner [ST] joint at opposite end
  • One o-ring included
  • Fabricated from borosilicate glass
Adapter features a #15 o-ring joint on one end with a Porosity B (70-100 micron) glass frit below the o-ring joint. An inner standard taper joint is at the opposite end. Supplied with one o-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty