Would you like to log in again?
Hose Connection Adapter, Bent, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5205-25 5205-16 5205-05 5205-10 5205-114 5205-35 5205-120 5205-15 9088-07 5205-110 5205-124 5205-112 9088-09
89082-630EA 72.44 USD InStock
89082-630 89064-866 89064-864 89051-402 89051-510 89051-434 89051-420 89051-430 10767-890 10098-738 10098-736 89082-634 89082-632
Hose Connection Adapter, Bent, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These Hose Connection Adapters are available with either a standard taper inner or a spherical ball bottom joint.
  • Inner [ST] or spherical ball bottom joint
  • 90° angle top hose connection
  • Fabricated from borosilicate glass
At the top of the adapter is a bent 90° hose connection. Fabricated from borosilicate glass. Also available with an integral Porosity A (145–174 micron) fritted disc.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty