Would you like to log in again?
Adapter, Septum Inlet, Ace Glass Incorporated
Supplier: Ace Glass
5113-13 5113-23
89082-706EA 71.48 USD InStock
89082-706 89082-704
Adapter, Septum Inlet, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This is a sampling adapter with a thread at top that accepts a cap with hole and a PTFE faced septum to allow insertion of a syringe needle.
  • Bottom [ST] inner joint
  • Threaded top
  • Accommodates syringe needle
  • Cap and septum included
  • Fabricated from borosilicate glass
Adapter features a standard taper inner joint at the bottom and an 8-425 GPI thread at the top that accepts an 89075-244 cap with hole and an 89064-072 PTFE faced septum to allow insertion of a syringe needle. Supplied complete with cap and septum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty